siRNA Details
| Gene Symbol: Chka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017127.1 | NP_058823.1 | 4 | 852 | ||
| XM_006230707.2 | XP_006230769.1 | 6 | 1875 | |||
| XM_006230708.2 | XP_006230770.1 | 5 | 1844 | |||
| XM_008760090.1 | XP_008758312.1 | 4 | 3167 | |||
| XM_008760091.1 | XP_008758313.1 | 5 | 3327 | |||
| GenBank mRNA | BC081821.1 | - | 4 | 828 | 7109261 | |
| D10262.1 | - | 4 | 852 | |||
| D37885.1 | - | 5 | 608 | |||

Chka
Genomic Map