siRNA Details
Gene Symbol: Chad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019164.1 | NP_062037.1 | 1 | 667 | ||
XM_006247186.1 | XP_006247248.1 | 1 | 2745 | |||
GenBank mRNA | AF004953.1 | - | 1 | 667 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019164.1 | NP_062037.1 | 1 | 667 | ||
XM_006247186.1 | XP_006247248.1 | 1 | 2745 | |||
GenBank mRNA | AF004953.1 | - | 1 | 667 |