siRNA Details
| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019165.1 | NP_062038.1 | 4, 5 | 368 | ||
| GenBank mRNA | AJ222813.1 | - | 4, 5 | 368 | ||
| AY077842.1 | - | 596 | ||||
| AY258448.1 | - | 4, 5 | 368 | |||
| U77776.1 | - | 4, 5 | 368 | |||

Il18
Genomic Map