siRNA Details
| Gene Symbol: Hfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053301.4 | NP_445753.1 | 2 | 185 | ||
| GenBank mRNA | AJ001517.1 | - | 2 | 224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hfe| Gene Symbol: Hfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053301.4 | NP_445753.1 | 2 | 185 | ||
| GenBank mRNA | AJ001517.1 | - | 2 | 224 | ||