siRNA Details
| Gene Symbol: Ctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017129.1 | NP_058825.1 | 2 | 146 | ||
| XM_006230221.2 | XP_006230283.1 | 2 | 909 | |||
| GenBank mRNA | BC072690.1 | - | 2 | 135 | 7096800 | |
| D78591.1 | - | 2 | 146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctf1| Gene Symbol: Ctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017129.1 | NP_058825.1 | 2 | 146 | ||
| XM_006230221.2 | XP_006230283.1 | 2 | 909 | |||
| GenBank mRNA | BC072690.1 | - | 2 | 135 | 7096800 | |
| D78591.1 | - | 2 | 146 | |||