siRNA Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017130.2 | NP_058826.2 | 3 | 932 | ||
| XM_006245361.2 | XP_006245423.1 | 3 | 872 | |||
| XM_006245362.2 | XP_006245424.1 | 3 | 1536 | |||
| XM_006245363.2 | XP_006245425.1 | 3 | 883 | |||
| XM_006245364.1 | XP_006245426.1 | 3 | 838 | |||
| XM_006245365.2 | XP_006245427.1 | 3 | 879 | |||
| XM_008767209.1 | XP_008765431.1 | 4 | 1064 | |||
| XM_008767210.1 | XP_008765432.1 | 4 | 1010 | |||
| XM_008767211.1 | XP_008765433.1 | 3 | 1018 | |||
| GenBank mRNA | BC091154.1 | - | 3 | 861 | 7323285 | |
| D16300.1 | - | 3 | 932 | |||
| FQ213221.1 | - | 3 | 895 | |||
| FQ216178.1 | - | 3 | 807 | |||

Neu2
Genomic Map