siRNA Details
| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019166.2 | NP_062039.1 | 4 | 561 | ||
| GenBank mRNA | U39549.1 | - | 4 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syngr1| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019166.2 | NP_062039.1 | 4 | 561 | ||
| GenBank mRNA | U39549.1 | - | 4 | 530 | ||