siRNA Details
| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031564.1 | NP_113752.1 | 1, 2 | 123 | ||
| XM_006247976.2 | XP_006248038.1 | 1, 2 | 207 | |||
| GenBank mRNA | U69278.1 | - | 1, 2 | 123 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha3| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031564.1 | NP_113752.1 | 1, 2 | 123 | ||
| XM_006247976.2 | XP_006248038.1 | 1, 2 | 207 | |||
| GenBank mRNA | U69278.1 | - | 1, 2 | 123 | ||