siRNA Details
| Gene Symbol: Tubb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080882.1 | NP_543158.1 | 4 | 714 | ||
| GenBank mRNA | BC168216.1 | - | 4 | 719 | 7315263 | |
| DY315612.1 | - | 1 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubb4a| Gene Symbol: Tubb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080882.1 | NP_543158.1 | 4 | 714 | ||
| GenBank mRNA | BC168216.1 | - | 4 | 719 | 7315263 | |
| DY315612.1 | - | 1 | 275 | |||