siRNA Details
| Gene Symbol: Arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019168.1 | NP_062041.1 | 6 | 768 | ||
| GenBank mRNA | U90887.1 | - | 6 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arg2| Gene Symbol: Arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019168.1 | NP_062041.1 | 6 | 768 | ||
| GenBank mRNA | U90887.1 | - | 6 | 768 | ||