siRNA Details
| Gene Symbol: Rcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017132.2 | NP_058828.2 | 4 | 726 | ||
| GenBank mRNA | BC061962.1 | - | 4 | 680 | 5598390 | |
| U15734.1 | - | 4 | 737 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcn2| Gene Symbol: Rcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017132.2 | NP_058828.2 | 4 | 726 | ||
| GenBank mRNA | BC061962.1 | - | 4 | 680 | 5598390 | |
| U15734.1 | - | 4 | 737 | |||