siRNA Details
| Gene Symbol: Snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019169.2 | NP_062042.1 | 3 | 307 | ||
| XM_006236591.2 | XP_006236653.1 | 3 | 588 | |||
| GenBank mRNA | AF007758.1 | - | 2 | 170 | ||
| AY550005.1 | - | 2 | 170 | |||
| AY550006.1 | - | 2 | 170 | |||
| BC087682.1 | - | 3 | 307 | 7321273 | ||
| FQ212580.1 | - | 3 | 284 | |||
| FQ227096.1 | - | 3 | 298 | |||
| S73007.1 | - | 3 | 238 | |||
| S73008.1 | - | 3 | 238 | |||

Snca
Genomic Map