siRNA Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017134.3 | NP_058830.2 | 7 | 903 | ||
| GenBank mRNA | BC091158.1 | - | 7 | 833 | 7368991 | |
| FM120421.1 | - | 5 | 545 | |||
| FQ209619.1 | - | 4 | 414 | |||
| FQ219159.1 | - | 7 | 785 | |||
| J02720.1 | - | 7 | 784 | |||

Arg1
Genomic Map