siRNA Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017134.3 | NP_058830.2 | 8 | 1066 | ||
| GenBank mRNA | BC091158.1 | - | 8 | 996 | 7368991 | |
| FQ209619.1 | - | 5 | 577 | |||
| FQ218591.1 | - | 8 | 1015 | |||
| FQ219159.1 | - | 8 | 948 | |||
| J02720.1 | - | 8 | 947 | |||

Arg1
Genomic Map