siRNA Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017134.3 | NP_058830.2 | 4 | 512 | ||
| GenBank mRNA | BC091158.1 | - | 4 | 442 | 7368991 | |
| FM100195.1 | - | 4 | 481 | |||
| FM120421.1 | - | 2 | 154 | |||
| FQ209619.1 | - | 1 | 22 | |||
| FQ218591.1 | - | 4 | 456 | |||
| FQ219159.1 | - | 4 | 393 | |||
| J02720.1 | - | 4 | 393 | |||

Arg1
Genomic Map