siRNA Details
| Gene Symbol: Ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017135.3 | NP_058831.1 | 4 | 518 | ||
| XM_006238445.2 | XP_006238507.1 | 5 | 823 | |||
| GenBank mRNA | BC087024.1 | - | 4 | 570 | 7107348 | |
| D87809.1 | - | 4 | 518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak4| Gene Symbol: Ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017135.3 | NP_058831.1 | 4 | 518 | ||
| XM_006238445.2 | XP_006238507.1 | 5 | 823 | |||
| GenBank mRNA | BC087024.1 | - | 4 | 570 | 7107348 | |
| D87809.1 | - | 4 | 518 | |||