siRNA Details
| Gene Symbol: Nfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031567.1 | NP_113755.1 | 3 | 625 | ||
| XM_006240850.2 | XP_006240912.1 | 3 | 711 | |||
| GenBank mRNA | AB012233.2 | - | 3 | 625 | ||
| FQ213370.1 | - | 3 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfic| Gene Symbol: Nfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031567.1 | NP_113755.1 | 3 | 625 | ||
| XM_006240850.2 | XP_006240912.1 | 3 | 711 | |||
| GenBank mRNA | AB012233.2 | - | 3 | 625 | ||
| FQ213370.1 | - | 3 | 677 | |||