siRNA Details
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017136.2 | NP_058832.1 | 2 | 509 | ||
GenBank mRNA | BC097330.1 | - | 2 | 509 | 7460860 | |
D37920.1 | - | 2 | 640 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017136.2 | NP_058832.1 | 2 | 509 | ||
GenBank mRNA | BC097330.1 | - | 2 | 509 | 7460860 | |
D37920.1 | - | 2 | 640 |