siRNA Details
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031568.2 | NP_113756.1 | 14 | 1262 | ||
GenBank mRNA | Z67744.1 | - | 14 | 1263 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031568.2 | NP_113756.1 | 14 | 1262 | ||
GenBank mRNA | Z67744.1 | - | 14 | 1263 |