siRNA Details
| Gene Symbol: Galr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019172.4 | NP_062045.1 | 2 | 597 | ||
| GenBank mRNA | AF008548.1 | - | 2 | 718 | ||
| AF010318.1 | - | 2 | 597 | |||
| U94322.1 | - | 2 | 1067 | |||
| Y15248.1 | - | 2 | 577 | |||
| Gene Symbol: LOC103690156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768432.1 | XP_008766654.1 | 1619 | |||
More Information
Set Membership: |
|

Galr2
Genomic Map