siRNA Details
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017138.2 | NP_058834.1 | 4, 5 | 560 | ||
XM_008766672.1 | XP_008764894.1 | 4, 5 | 633 | |||
GenBank mRNA | BC060578.1 | - | 4, 5 | 578 | 6919235 | |
D25224.1 | - | 4, 5 | 561 | |||
FJ386454.1 | - | 3, 4 | 541 | |||
FQ209463.1 | - | 4, 5 | 574 | |||
FQ209490.1 | - | 4, 5 | 575 | |||
FQ209667.1 | - | 4, 5 | 574 | |||
FQ209729.1 | - | 4, 5 | 574 | |||
FQ211189.1 | - | 4, 5 | 574 | |||
FQ212660.1 | - | 4, 5 | 574 | |||
FQ212893.1 | - | 4, 5 | 574 | |||
FQ213317.1 | - | 4, 5 | 573 | |||
FQ213334.1 | - | 4, 5 | 574 | |||
FQ213623.1 | - | 4, 5 | 577 | |||
FQ213835.1 | - | 4, 5 | 575 | |||
FQ213894.1 | - | 4, 5 | 574 | |||
FQ215242.1 | - | 4, 5 | 574 | |||
FQ215292.1 | - | 4, 5 | 575 | |||
FQ216085.1 | - | 4, 5 | 574 | |||
FQ216130.1 | - | 4, 5 | 574 | |||
FQ216245.1 | - | 4, 5 | 574 | |||
FQ216788.1 | - | 4, 5 | 576 | |||
FQ216997.1 | - | 4, 5 | 574 | |||
FQ217664.1 | - | 4, 5 | 574 | |||
FQ219586.1 | - | 4, 5 | 574 | |||
FQ219785.1 | - | 4, 5 | 577 | |||
FQ220012.1 | - | 4, 5 | 574 | |||
FQ220017.1 | - | 4, 5 | 572 | |||
FQ220075.1 | - | 4, 5 | 575 | |||
FQ220084.1 | - | 4, 5 | 577 | |||
FQ220126.1 | - | 4, 5 | 574 | |||
FQ220134.1 | - | 4, 5 | 574 | |||
FQ220171.1 | - | 4, 5 | 573 | |||
FQ220184.1 | - | 4, 5 | 574 | |||
FQ220347.1 | - | 4, 5 | 576 | |||
FQ220461.1 | - | 4, 5 | 555 | |||
FQ220468.1 | - | 4, 5 | 574 | |||
FQ220661.1 | - | 4, 5 | 577 | |||
FQ220822.1 | - | 4, 5 | 574 | |||
FQ220904.1 | - | 4, 5 | 575 | |||
FQ220964.1 | - | 4, 5 | 574 | |||
FQ221103.1 | - | 4, 5 | 574 | |||
FQ221334.1 | - | 4, 5 | 594 | |||
FQ221443.1 | - | 4, 5 | 575 | |||
FQ221672.1 | - | 4, 5 | 574 | |||
FQ222093.1 | - | 4, 5 | 576 | |||
FQ222280.1 | - | 4, 5 | 574 | |||
FQ222542.1 | - | 4, 5 | 576 | |||
FQ222757.1 | - | 4, 5 | 572 | |||
FQ223223.1 | - | 4, 5 | 574 | |||
FQ224747.1 | - | 4, 5 | 574 | |||
FQ225510.1 | - | 4, 5 | 575 | |||
FQ226054.1 | - | 4, 5 | 574 | |||
FQ226408.1 | - | 4, 5 | 574 | |||
FQ227056.1 | - | 4, 5 | 574 | |||
FQ227293.1 | - | 4, 5 | 574 | |||
FQ228002.1 | - | 4, 5 | 574 | |||
FQ228212.1 | - | 4, 5 | 575 | |||
FQ229290.1 | - | 4, 5 | 576 | |||
FQ229305.1 | - | 4, 5 | 576 | |||
FQ229367.1 | - | 4, 5 | 575 | |||
FQ229516.1 | - | 4, 5 | 574 | |||
FQ229579.1 | - | 4, 5 | 574 | |||
FQ229598.1 | - | 4, 5 | 575 | |||
FQ229712.1 | - | 4, 5 | 574 | |||
FQ229787.1 | - | 4, 5 | 574 | |||
FQ229850.1 | - | 4, 5 | 576 | |||
FQ229859.1 | - | 4, 5 | 572 | |||
FQ229890.1 | - | 4, 5 | 574 | |||
FQ229918.1 | - | 4, 5 | 594 | |||
FQ229945.1 | - | 4, 5 | 574 | |||
FQ229970.1 | - | 4, 5 | 574 | |||
FQ230088.1 | - | 4, 5 | 574 | |||
FQ230106.1 | - | 4, 5 | 574 | |||
FQ230258.1 | - | 4, 5 | 573 | |||
FQ231469.1 | - | 3, 4 | 391 | |||
FQ231622.1 | - | 4, 5 | 572 | |||
FQ232305.1 | - | 4, 5 | 576 | |||
FQ232568.1 | - | 4, 5 | 575 | |||
FQ233068.1 | - | 4, 5 | 574 | |||
FQ233112.1 | - | 4, 5 | 574 | |||
FQ233340.1 | - | 4, 5 | 576 | |||
FQ233547.1 | - | 4, 5 | 575 | |||
FQ233919.1 | - | 4, 5 | 574 | |||
FQ234511.1 | - | 4, 5 | 574 | |||
FQ234742.1 | - | 4, 5 | 574 | |||
FQ234765.1 | - | 4, 5 | 574 | |||
FQ234772.1 | - | 4, 5 | 574 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011029, AK011041, AK010985, AK088954, BC094902, and others.
Panther Classification:
Gene Ontology Categories:
