siRNA Details
| Gene Symbol: Drd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017140.1 | NP_058836.1 | 4 | 754 | ||
| XM_006248313.2 | XP_006248375.2 | 5 | 971 | |||
| GenBank mRNA | X53944.1 | - | 4 | 754 | ||
More Information
Other Species Targeted
Set Membership: |
|

Drd3
Genomic Map