siRNA Details
| Gene Symbol: Adcy8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017142.1 | NP_058838.1 | 10 | 3051 | ||
| XM_006241703.2 | XP_006241765.1 | 10 | 3544 | |||
| XM_006241704.2 | XP_006241766.1 | 9 | 3346 | |||
| GenBank mRNA | L26986.1 | - | 10 | 3051 | ||

Adcy8
Genomic Map