siRNA Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 5, 6 | 505 | ||
| GenBank mRNA | BC166508.1 | - | 5, 6 | 488 | ||
| D00688.1 | - | 5, 6 | 511 | |||
| FM037037.1 | - | 5, 6 | 455 | |||
| FM101032.1 | - | 5, 6 | 504 | |||
| FM117485.1 | - | 4, 5 | 381 | |||
| FM132632.1 | - | 1, 2 | 31 | |||

Maoa
Genomic Map