siRNA Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 7, 8 | 797 | ||
| GenBank mRNA | BC166508.1 | - | 7, 8 | 780 | ||
| D00688.1 | - | 7, 8 | 803 | |||
| FM132632.1 | - | 3, 4 | 323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maoa| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033653.1 | NP_387502.1 | 7, 8 | 797 | ||
| GenBank mRNA | BC166508.1 | - | 7, 8 | 780 | ||
| D00688.1 | - | 7, 8 | 803 | |||
| FM132632.1 | - | 3, 4 | 323 | |||