siRNA Details
| Gene Symbol: Mgll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138502.2 | NP_612511.1 | 2 | 204 | ||
| XM_008763053.1 | XP_008761275.1 | 2 | 862 | |||
| GenBank mRNA | AY081195.1 | - | 1 | 87 | ||
| BC107920.1 | - | 2 | 204 | 7446100 | ||
| FQ227013.1 | - | 2 | 830 | |||

Mgll
Genomic Map