siRNA Details
| Gene Symbol: Sell | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019177.3 | NP_062050.3 | 3 | 298 | ||
| GenBank mRNA | D10831.1 | - | 3 | 290 | ||
| S79523.1 | - | 3 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sell| Gene Symbol: Sell | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019177.3 | NP_062050.3 | 3 | 298 | ||
| GenBank mRNA | D10831.1 | - | 3 | 290 | ||
| S79523.1 | - | 3 | 298 | |||