siRNA Details
| Gene Symbol: Mcpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017145.2 | NP_058841.1 | 4 | 561 | ||
| GenBank mRNA | AF063851.1 | - | 4 | 562 | ||
| FQ220114.1 | - | 4 | 561 | |||
| FQ220196.1 | - | 4 | 561 | |||
| FQ220424.1 | - | 4 | 523 | |||
| FQ220659.1 | - | 4 | 561 | |||
| FQ220808.1 | - | 4 | 594 | |||
| FQ221313.1 | - | 4 | 561 | |||
| FQ228378.1 | - | 4 | 561 | |||
| FQ229265.1 | - | 4 | 561 | |||
| FQ229320.1 | - | 4 | 561 | |||

Mcpt1
Genomic Map