siRNA Details
| Gene Symbol: Cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017147.2 | NP_058843.1 | 2 | 247 | ||
| GenBank mRNA | BC059143.1 | - | 2 | 247 | 6889053 | |
| BC086533.1 | - | 2 | 232 | 7305422 | ||
| FQ212136.1 | - | 1 | 120 | |||
| FQ212794.1 | - | 2 | 246 | |||
| FQ212910.1 | - | 2 | 242 | |||
| FQ213701.1 | - | 2 | 242 | |||
| FQ213897.1 | - | 2 | 271 | |||
| FQ214120.1 | - | 2 | 243 | |||
| FQ214438.1 | - | 2 | 242 | |||
| FQ220384.1 | - | 2 | 258 | |||
| FQ220571.1 | - | 2 | 244 | |||
| FQ226405.1 | - | 2 | 242 | |||
| FQ227487.1 | - | 2 | 244 | |||
| FQ229073.1 | - | 2 | 271 | |||
| FQ229766.1 | - | 2 | 246 | |||
| FQ229833.1 | - | 2 | 242 | |||
| FQ229875.1 | - | 2 | 246 | |||
| FQ230001.1 | - | 2 | 246 | |||
| FQ230091.1 | - | 2 | 243 | |||
| FQ231379.1 | - | 2 | 242 | |||
| FQ232577.1 | - | 1 | 122 | |||
| FQ233750.1 | - | 2 | 242 | |||
| FQ234162.1 | - | 2 | 246 | |||
| FQ234303.1 | - | 2 | 242 | |||
| X62908.1 | - | 1 | 171 | |||

Cfl1
Genomic Map