siRNA Details
| Gene Symbol: Actc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019183.1 | NP_062056.1 | 4 | 673 | ||
| GenBank mRNA | BC127451.1 | - | 4 | 548 | 7379877 | |
| CF109672.1 | - | 4 | 527 | |||
| CK356511.1 | - | 4 | 673 | |||
| X80130.1 | - | 4 | 559 | |||

Actc1
Genomic Map