siRNA Details
| Gene Symbol: Lasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032613.2 | NP_116002.1 | 7 | 722 | ||
| XM_006247288.2 | XP_006247350.1 | 7 | 654 | |||
| GenBank mRNA | AF242187.1 | - | 7 | 636 | ||
| BC099791.1 | - | 7 | 722 | 7934892 | ||

Lasp1
Genomic Map