siRNA Details
| Gene Symbol: Meox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017149.1 | NP_058845.1 | 2 | 857 | ||
| GenBank mRNA | Z17223.1 | - | 2 | 857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Meox2| Gene Symbol: Meox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017149.1 | NP_058845.1 | 2 | 857 | ||
| GenBank mRNA | Z17223.1 | - | 2 | 857 | ||