siRNA Details
| Gene Symbol: AMBN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016519.5 | NP_057603.1 | 5, 6 | 392 | ||
| GenBank mRNA | AF209780.1 | - | 5, 6 | 374 | ||
| AF219994.2 | - | 5, 6 | 356 | |||
| AF263464.1 | - | 5, 6 | 366 | |||
| AJ537437.1 | - | 5, 6 | 366 | |||
| AK314918.1 | - | 5, 6 | 327 | |||
| BC106931.1 | - | 5, 6 | 327 | 40033703 | ||
| BC106932.1 | - | 5, 6 | 327 | 40033705 | ||
| BP248666.1 | - | 5, 6 | 392 | |||

AMBN
Genomic Map