siRNA Details
| Gene Symbol: Hal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017159.1 | NP_058855.1 | 11 | 1050 | ||
| GenBank mRNA | BC089809.1 | - | 11 | 1079 | 7325816 | |
| M58308.1 | - | 11 | 1050 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hal| Gene Symbol: Hal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017159.1 | NP_058855.1 | 11 | 1050 | ||
| GenBank mRNA | BC089809.1 | - | 11 | 1079 | 7325816 | |
| M58308.1 | - | 11 | 1050 | |||