siRNA Details
| Gene Symbol: Adora2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017161.1 | NP_058857.1 | 2 | 708 | ||
| GenBank mRNA | BC161854.1 | - | 2 | 716 | 9024632 | |
| M91466.1 | - | 2 | 708 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adora2b| Gene Symbol: Adora2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017161.1 | NP_058857.1 | 2 | 708 | ||
| GenBank mRNA | BC161854.1 | - | 2 | 716 | 9024632 | |
| M91466.1 | - | 2 | 708 | |||
Set Membership: |
|