siRNA Details
| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177425.3 | NP_803174.2 | 5 | 536 | ||
| GenBank mRNA | BC086356.1 | - | 5 | 550 | 7303714 | |
| CD373038.1 | - | 5 | 527 | |||
| U44948.1 | - | 5 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp2| Gene Symbol: Csrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177425.3 | NP_803174.2 | 5 | 536 | ||
| GenBank mRNA | BC086356.1 | - | 5 | 550 | 7303714 | |
| CD373038.1 | - | 5 | 527 | |||
| U44948.1 | - | 5 | 541 | |||