siRNA Details
| Gene Symbol: Plcb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033350.1 | NP_203501.1 | 21 | 2533 | ||
| XM_006230711.2 | XP_006230773.1 | 21 | 2665 | |||
| GenBank mRNA | DQ120507.1 | - | 21 | 2533 | ||
| DQ120508.1 | - | 21 | 2533 | |||
| M99567.1 | - | 21 | 2467 | |||

Plcb3
Genomic Map