siRNA Details
| Gene Symbol: SLIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003061.2 | NP_003052.2 | 16, 17 | 1890 | ||
| GenBank mRNA | AB011537.3 | - | 16, 17 | 1890 | ||
| AB017167.1 | - | 16, 17 | 1870 | |||
| AY029183.1 | - | 16, 17 | 1617 | |||
| BC146761.1 | - | 16, 17 | 1890 | 9007200 | ||
| BC146851.1 | - | 16, 17 | 1803 | 9056779 | ||

SLIT1
Genomic Map