siRNA Details
| Gene Symbol: Serpinh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017173.1 | NP_058869.1 | 5 | 1126 | ||
| GenBank mRNA | BC086529.1 | - | 4 | 1017 | 7303866 | |
| M69246.1 | - | 5 | 1126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinh1| Gene Symbol: Serpinh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017173.1 | NP_058869.1 | 5 | 1126 | ||
| GenBank mRNA | BC086529.1 | - | 4 | 1017 | 7303866 | |
| M69246.1 | - | 5 | 1126 | |||