siRNA Details
| Gene Symbol: Faah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024132.3 | NP_077046.1 | 8 | 1056 | ||
| GenBank mRNA | BC091148.1 | - | 1057 | 7370226 | ||
| U72497.1 | - | 1044 | ||||
| Gene Symbol: LOC100911581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750001.3 | XP_003750049.1 | 8 | 1401 | ||

Faah
Genomic Map