siRNA Details
| Gene Symbol: Phkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031573.1 | NP_113761.1 | 3 | 217 | ||
| GenBank mRNA | X07320.1 | - | 3 | 217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phkg1| Gene Symbol: Phkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031573.1 | NP_113761.1 | 3 | 217 | ||
| GenBank mRNA | X07320.1 | - | 3 | 217 | ||