siRNA Details
| Gene Symbol: Pla2g5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017174.1 | NP_058870.1 | 3 | 324 | ||
| XM_006239139.2 | XP_006239201.1 | 4 | 1123 | |||
| XM_006239140.2 | XP_006239202.1 | 4 | 1702 | |||
| XM_006239141.2 | XP_006239203.1 | 3 | 246 | |||
| XM_008764209.1 | XP_008762431.1 | 5 | 1237 | |||
| XM_008764210.1 | XP_008762432.1 | 3 | 2344 | |||
| XM_008764211.1 | XP_008762433.1 | 2 | 359 | |||
| GenBank mRNA | AY651028.1 | - | 3 | 191 | ||
| BC085745.1 | - | 3 | 349 | 7192819 | ||
| U03763.1 | - | 3 | 324 | |||

Pla2g5
Genomic Map