siRNA Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083911.1 | NP_001077380.1 | 2 | 252 | ||
| NM_019192.2 | NP_062065.2 | 2 | 180 | |||
| GenBank mRNA | BC059137.1 | - | 2 | 181 | ||
| BC072539.1 | - | 2 | 169 | 7104195 | ||
| CB699091.1 | - | 2 | 252 | |||
| CB745388.1 | - | 2 | 404 | |||
| D25221.1 | - | 2 | 161 | |||
| DN933732.1 | - | 2 | 500 | |||
| M63574.1 | - | 140 | ||||

Sepp1
Genomic Map