siRNA Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003063.2 | NP_003054.1 | 2 | 229 | ||
| GenBank mRNA | AK312097.1 | - | 2 | 224 | ||
| BC005261.1 | - | 2 | 224 | 3934241 | ||
| BC094685.1 | - | 2 | 211 | 6203008 | ||
| BC104150.2 | - | 2 | 181 | 40029313 | ||
| BC104185.2 | - | 2 | 176 | 40030832 | ||
| BC113930.1 | - | 2 | 180 | 40030834 | ||
| BC113987.1 | - | 2 | 158 | 40030833 | ||
| BP233121.1 | - | 2 | 229 | |||
| CR450290.1 | - | 1 | 55 | |||
| U96094.1 | - | 2 | 223 | |||

SLN
Genomic Map