siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019195.2 | NP_062068.1 | 2 | 175 | ||
| XM_006248262.2 | XP_006248324.1 | 2 | 754 | |||
| XM_006248263.2 | XP_006248325.1 | 2 | 754 | |||
| XM_006248264.2 | XP_006248326.1 | 2 | 754 | |||
| XM_006248265.2 | XP_006248327.1 | 2 | 754 | |||
| XM_006248266.2 | XP_006248328.1 | 2 | 197 | |||
| XM_006248267.2 | XP_006248329.1 | 2 | 754 | |||
| XM_006248268.2 | XP_006248330.1 | 2 | 952 | |||
| GenBank mRNA | AF017437.1 | - | 2 | 61 | ||
| BC085740.1 | - | 2 | 175 | 7189894 | ||
| D87659.1 | - | 2 | 85 | |||

Cd47
Genomic Map