siRNA Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019197.1 | NP_062070.1 | 3 | 518 | ||
| XM_006245161.2 | XP_006245223.1 | 3 | 1181 | |||
| XM_006245162.2 | XP_006245224.1 | 4 | 645 | |||
| XM_008767212.1 | XP_008765434.1 | 3 | 505 | |||
| GenBank mRNA | AJ007480.1 | - | 1 | 75 | ||
| AY780673.1 | - | 2 | 394 | |||
| CK357674.1 | - | 3 | 518 | |||
| FQ228974.1 | - | 1 | 87 | |||
| M17784.1 | - | 2 | 394 | |||

Serpine2
Genomic Map