siRNA Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019197.1 | NP_062070.1 | 4, 5 | 811 | ||
| XM_006245161.2 | XP_006245223.1 | 4, 5 | 1474 | |||
| XM_006245162.2 | XP_006245224.1 | 5, 6 | 938 | |||
| XM_008767212.1 | XP_008765434.1 | 4, 5 | 798 | |||
| GenBank mRNA | AY780673.1 | - | 3, 4 | 686 | ||
| FQ228974.1 | - | 2, 3 | 380 | |||
| M17784.1 | - | 3, 4 | 687 | |||

Serpine2
Genomic Map