siRNA Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019197.1 | NP_062070.1 | 4 | 654 | ||
| XM_006245161.2 | XP_006245223.1 | 4 | 1317 | |||
| XM_006245162.2 | XP_006245224.1 | 5 | 781 | |||
| XM_008767212.1 | XP_008765434.1 | 4 | 641 | |||
| GenBank mRNA | AY780673.1 | - | 3 | 529 | ||
| CK357674.1 | - | 4 | 654 | |||
| M17784.1 | - | 3 | 530 | |||

Serpine2
Genomic Map