siRNA Details
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182814.2 | NP_877966.1 | 6, 7 | 778 | ||
GenBank mRNA | AY223861.1 | - | 6, 7 | 644 | ||
BC079283.1 | - | 6, 7 | 749 | 7134210 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182814.2 | NP_877966.1 | 6, 7 | 778 | ||
GenBank mRNA | AY223861.1 | - | 6, 7 | 644 | ||
BC079283.1 | - | 6, 7 | 749 | 7134210 |