siRNA Details
| Gene Symbol: Phlda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017180.2 | NP_058876.2 | 1 | 815 | ||
| GenBank mRNA | AF192802.1 | - | 1 | 793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phlda1| Gene Symbol: Phlda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017180.2 | NP_058876.2 | 1 | 815 | ||
| GenBank mRNA | AF192802.1 | - | 1 | 793 | ||